A10492.36
A10492.22
A10492.14
AB116343.0100
A10492.36EA
253
EUR
InStock
A10492.36
A10492.22
A10492.14
ABCRAB116343.0100
Diphenylsilandiol ≥98%
Diphenylsilandiol
Purity: 98%
CAS No.: 947-42-2
Molecular Formula: C12H12O2Si
Formula Weight: 216.31
Odour: Odourless
Melting Point: ca 150° dec.
Beilstein Reference: 2523445
UN No.: 1325
MDL No.: MFCD00002101