A10514.09
A10514.18
A10514.09EA
55.7
EUR
InStock
A10514.09
A10514.18
3,5-Di-tert-butyl-4-hydroxycinnamic acid überwiegend trans 98%
3,5-Di-tert-butyl-4-hydroxycinnamic acid
Purity: 98%
CAS No.: 2243-42-7
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 111-115°
Beilstein Reference: 978452