A10561.14
A10561.14EA
40.8
EUR
InStock
A10561.14
N,N,2,4,6-Pentamethylanilin ≥98%
N,N,2,4,6-Pentamethylanilin
Purity: 98%
CAS No.: 13021-15-3
Molecular Formula: C11H17N
Formula Weight: 163.27
Flash Point: 78°(174°F)
Refractive Index: 1.5210
Boiling Point: 213-215°
MDL No.: MFCD00010596