A10577.14
A10577.22
A10577.36
A10577.14EA
34
EUR
InStock
A10577.14
A10577.22
A10577.36
1-Bromnonan ≥99%
1-Bromnonan
Purity: 99%
CAS No.: 693-58-3
Molecular Formula: CH3(CH2)8Br
Formula Weight: 207.16
Flash Point: 90°(194°F)
Refractive Index: 1.4550
Boiling Point: 200-201°
Beilstein Reference: 1733632
MDL No.: MFCD00000278