A10609.09
A10609.18
A10609.30
A10609.09EA
25.3
EUR
InStock
A10609.09
A10609.18
A10609.30
2,3,4-Trimethoxybenzaldehyd ≥98%
2,3,4-Trimethoxybenzaldehyde
Purity: 98+%
CAS No.: 2103-57-3
Molecular Formula: C10H12O4
Formula Weight: 196.20
Flash Point: 110°(230°F)
Refractive Index: 1.5547
Melting Point: 38-42°
Boiling Point: 168-170°/12mm
Beilstein Reference: 981091
MDL No.: MFCD00003310