A10643.0E
A10643.22
A10643.36
A10643.0EEA
678
EUR
InStock
A10643.0E
A10643.22
A10643.36
D(+)-Xylose ≥98%
D(+)-Xylose
Purity: 98+%
CAS No.: 58-86-6
Molecular Formula: C5H10O5
Formula Weight: 150.13
Flash Point: 100°(212°F)
Physical Form: Cryst powder
Density: 1.525
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 147-151°
Beilstein Reference: 1562108
Merck Reference: 14,10087
Optical Rotation: +19° (c=10 in water + NH4OH)
MDL No.: MFCD00151475