A10648.03
A10648.06
A10648.14
A10648.03EA
25.3
EUR
InStock
A10648.03
A10648.06
A10648.14
3-Methoxythiophenol ≥98%
3-Methoxythiophenol
Purity: 98%
CAS No.: 15570-12-4
Molecular Formula: C7H8OS
Formula Weight: 140.21
Flash Point: 96°(204°F)
Refractive Index: 1.5865
Odour: Unpleasant
Boiling Point: 225-227°
Beilstein Reference: 2041496
MDL No.: MFCD00004841