A10776.0C
A10776.0E
A10776.36
A10776.0CEA
492
EUR
InStock
A10776.0C
A10776.0E
A10776.36
Pivalinsäure ≥99%
Pivalinsäure
Purity: 99%
CAS No.: 75-98-9
Molecular Formula: (CH3)3CCO2H
Formula Weight: 102.13
Flash Point: 63°(145°F)
Odour: Pungent
Boiling Point: 163-165°
Solubility: Soluble in alcohol, ether. Slightly soluble in water
Beilstein Reference: 969480
Merck Reference: 14,7511
UN No.: 3261
MDL No.: MFCD00004194