A10901.30
A10901.0B
A10901.18
A10901.30EA
47.3
EUR
InStock
A10901.30
A10901.0B
A10901.18
Caprylaldehyd ≥98%
Caprylaldehyd
Purity: 98%
CAS No.: 124-13-0
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 56°(133°F)
Refractive Index: 1.4195
Melting Point: 12-15°
Boiling Point: 169-171°
Merck Reference: 14,1766
UN No.: 1191
MDL No.: MFCD00007029