A10971.03
A10971.06
A10971.14
A10971.03EA
21.8
EUR
InStock
A10971.03
A10971.06
A10971.14
(4-Hydroxyphenyl)acetonitril ≥97%
(4-Hydroxyphenyl)acetonitril
Purity: 97%
CAS No.: 14191-95-8
Molecular Formula: C8H7NO
Formula Weight: 133.15
Melting Point: 66-71°
Boiling Point: 329-331°
MDL No.: MFCD00002383