A10997.0B
A10997.0I
A10997.30
A10997.36
A10997.0BEA
112
EUR
InStock
A10997.0B
A10997.0I
A10997.30
A10997.36
Benzylamin ≥98%
Benzylamin
Purity: 98+%
CAS No.: 100-46-9
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 72°(162°F)
Refractive Index: 1.5435
Melting Point: -43°
Boiling Point: 184-185°
Beilstein Reference: 741984
Merck Reference: 14,1125
UN No.: 2735
MDL No.: MFCD00008106