A11014.01
A11014.01EA
59.3
EUR
InStock
A11014.01
Bis(pentafluorphenyl)phenylphosphin ≥97%
Bis(pentafluorphenyl)phenylphosphin
Purity: 97%
CAS No.: 5074-71-5
Molecular Formula: C18H5F10P
Formula Weight: 442.20
Flash Point: 110°(230°F)
Melting Point: 59-64°
Boiling Point: 105°/0.3mm
Beilstein Reference: 6253897
MDL No.: MFCD00000291