A11123.0E
A11123.22
A11123.36
A11123.0EEA
221
EUR
InStock
A11123.0E
A11123.22
A11123.36
Antimon(III)oxid ≥99%
Antimon(III)oxid
Purity: 99%
CAS No.: 1309-64-4
Molecular Formula: Sb2O3
Formula Weight: 291.50
Odour: Odourless
Melting Point: 656°
Boiling Point: 1550° subl.
Merck Reference: 14,711
MDL No.: MFCD00011214