A11135.0D
A11135.0F
A11135.AP
A11135.0DEA
314
EUR
InStock
A11135.0D
A11135.0F
A11135.AP
tert-Butylamin 98%
tert-Butylamin
Purity: 98%
CAS No.: 75-64-9
Molecular Formula: (CH3)3CNH2
Formula Weight: 73.14
Flash Point: -38°(-36°F)
Refractive Index: 1.3780
Odour: Amine-like
Melting Point: -67°
Boiling Point: 44-46°
Beilstein Reference: 605267
Merck Reference: 14,1545
UN No.: 2733
MDL No.: MFCD00008050