A11141.0B
A11141.18
A11141.22
A11141.30
A11141.0BEA
294
EUR
InStock
A11141.0B
A11141.18
A11141.22
A11141.30
L(-)-Tyrosin ≥99%
L(-)-Tyrosin
Purity: 99%
CAS No.: 60-18-4
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Physical Form: Cryst powder
Density: 1.46
Appearance: White
Melting Point: 300°
Beilstein Reference: 392441
Merck Reference: 14,9839
Optical Rotation: -11° (c=4 in 1N HCl)
MDL No.: MFCD00002606