A11151.0E
A11151.30
A11151.36
A11151.0EEA
247
EUR
InStock
A11151.0E
A11151.30
A11151.36
2,5-Dichloranilin ≥99%
2,5-Dichloranilin
Purity: 99%
CAS No.: 95-82-9
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 139°(282°F)
Melting Point: 48-52°
Boiling Point: 250-251°
Beilstein Reference: 1447438
UN No.: 3442
MDL No.: MFCD00007667