A11249.09
A11249.18
A11249.30
A11249.09EA
51.7
EUR
InStock
A11249.09
A11249.18
A11249.30
18-Krone-6 (1,4,7,10,13,16-Hexaoxacyclooctadecan) ≥99%
18-Krone-6 (1,4,7,10,13,16-Hexaoxacyclooctadecan)
Purity: 99%
CAS No.: 17455-13-9
Molecular Formula: C12H24O6
Formula Weight: 264.32
Flash Point: 109°(228°F)
Odour: Odourless
Boiling Point: 116°/0.2mm
Beilstein Reference: 1619616
Merck Reference: 14,2602
MDL No.: MFCD00005113