A11295.0C
A11295.0E
A11295.36
A11295.0CEA
209
EUR
InStock
A11295.0C
A11295.0E
A11295.36
Melamin 99%
Melamin
Purity: 99%
CAS No.: 108-78-1
Molecular Formula: C3H6N6
Formula Weight: 126.12
Melting Point: ca 350° dec.
Beilstein Reference: 124341
Merck Reference: 14,5811
MDL No.: MFCD00006055