A11308.0E
A11308.22
A11308.36
A11308.0EEA
735
EUR
InStock
A11308.0E
A11308.22
A11308.36
Kaliumperiodat ≥99%
Kaliumperiodat
Purity: 99%
CAS No.: 7790-21-8
Molecular Formula: KIO4
Formula Weight: 230.00
Melting Point: 582°
Merck Reference: 14,7654
MDL No.: MFCD00011407