A11348.03
A11348.06
A11348.09
A11348.03EA
15.1
EUR
InStock
A11348.03
A11348.06
A11348.09
5-Chlor-2-thiophensulfonylchlorid ≥97%
5-Chlor-2-thiophensulfonylchlorid
Purity: 97%
CAS No.: 2766-74-7
Molecular Formula: C4H2Cl2O2S2
Formula Weight: 217.09
Storage Temperature: 2°C to 8°C
Flash Point: 110°(230°F)
Refractive Index: 1.5860
Boiling Point: 117-120°/14mm
Beilstein Reference: 130710
UN No.: 3265
MDL No.: MFCD00051667