A11377.18
A11377.30
A11377.18EA
41.5
EUR
InStock
A11377.18
A11377.30
4-Chloranisol ≥99%
4-Chloranisol
Purity: 99%
CAS No.: 623-12-1
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Flash Point: 78°(172°F)
Refractive Index: 1.5350
Melting Point: -18°
Boiling Point: 198-200°
Beilstein Reference: 1858901
MDL No.: MFCD00000616