A11427.09
A11427.18
A11427.09EA
47
EUR
InStock
A11427.09
A11427.18
(4-Bromphenyl)methylsulfon ≥98%
(4-Bromphenyl)methylsulfon
Purity: 98+%
CAS No.: 3466-32-8
Molecular Formula: C7H7BrO2S
Formula Weight: 235.10
Melting Point: 101-102°
Beilstein Reference: 2045675