A11439.14
A11439.22
A11439.36
A11439.14EA
34.1
EUR
InStock
A11439.14
A11439.22
A11439.36
1,2-Dihydroxybenzol-3,5-disulfonsäure Dinatriumsalz Monohydrat ≥97%
1,2-Dihydroxybenzol-3,5-disulfonsäure Dinatriumsalz Monohydrat
Purity: 97%
CAS No.: 270573-71-2
Molecular Formula: C6H4Na2O8S2.H2O
Formula Weight: 332.23 (314.21anhy)
Melting Point: 300°
Beilstein Reference: 4629740
Merck Reference: 14,9465
MDL No.: MFCD00007473