A11450.0E
A11450.22
A11450.36
A11450.0EEA
872
EUR
InStock
A11450.0E
A11450.22
A11450.36
Kaliumxanthogenat (Kalium-O-ethyldithiocarbonat) ≥97%
Kaliumxanthogenat (Kalium-O-ethyldithiocarbonat)
Purity: 98%
CAS No.: 140-89-6
Molecular Formula: C3H5KOS2
Formula Weight: 160.30
Flash Point: 205°(401°F)
Physical Form: Powder
Density: 1.558
Appearance: Yellow
Odour: Unpleasant
pH: 9-10
Melting Point: ca 205° dec.
Beilstein Reference: 3596974
Merck Reference: 14,7696
MDL No.: MFCD00004931