A11451.14
A11451.22
A11451.14EA
34.8
EUR
InStock
A11451.14
A11451.22
2-Amino-5-mercapto-1,3,4-thiadiazol 98%
2-Amino-5-mercapto-1,3,4-thiadiazole
Purity: 98+%
CAS No.: 2349-67-9
Molecular Formula: C2H3N3S2
Formula Weight: 133.19
Melting Point: ca 243° dec.
Beilstein Reference: 81728