A11488.0E
A11488.22
A11488.36
A11488.0EEA
461
EUR
InStock
A11488.0E
A11488.22
A11488.36
4-Chlor-3-nitrobenzotrifluorid ≥98%
4-Chlor-3-nitrobenzotrifluorid
Purity: 98%
CAS No.: 121-17-5
Molecular Formula: C7H3ClF3NO2
Formula Weight: 225.55
Flash Point: 101°(213°F)
Refractive Index: 1.4900
Melting Point: -3°
Boiling Point: 221-223°
Beilstein Reference: 523986
UN No.: 2307
MDL No.: MFCD00007084