A11516.03
A11516.06
A11516.14
A11516.03EA
65.9
EUR
InStock
A11516.03
A11516.06
A11516.14
1,4,8,11-Tetraazacyclotetradecan ≥98%
1,4,8,11-Tetraazacyclotetradecane
Purity: 98+%
CAS No.: 295-37-4
Molecular Formula: C10H24N4
Formula Weight: 200.33
Melting Point: 184-188°
Beilstein Reference: 111811