A11593.0E
A11593.22
A11593.36
A11593.0EEA
223
EUR
InStock
A11593.0E
A11593.22
A11593.36
Bromcyclohexan ≥98%
Bromcyclohexan
Purity: 98%
CAS No.: 108-85-0
Molecular Formula: C6H11Br
Formula Weight: 163.06
Flash Point: 62°(143°F)
Refractive Index: 1.4950
Melting Point: -57°
Boiling Point: 166-167°
Beilstein Reference: 1098373
Merck Reference: 14,2730
MDL No.: MFCD00003819