A11596.06
A11596.22
A11596.36
A11596.06EA
29
EUR
InStock
A11596.06
A11596.22
A11596.36
Phenylisothiocyanat ≥97%
Phenylisothiocyanat
Purity: 97%
CAS No.: 103-72-0
Molecular Formula: C7H5NS
Formula Weight: 135.19
Flash Point: 87°(188°F)
Refractive Index: 1.6500
Odour: Pungent
Melting Point: -21°
Boiling Point: 220-221°
Beilstein Reference: 471392
Merck Reference: 14,7297
UN No.: 2927
MDL No.: MFCD00004798