A11607.09
A11607.18
A11607.30
A11607.09EA
48.4
EUR
InStock
A11607.09
A11607.18
A11607.30
2-Pentylfuran ≥98%
2-Pentylfuran
Purity: 98%
CAS No.: 3777-69-3
Molecular Formula: C9H14O
Formula Weight: 138.21
Storage Temperature: 2°C to 8°C
Flash Point: 45°(113°F)
Refractive Index: 1.4500
Boiling Point: 167°
Beilstein Reference: 107854
UN No.: 1993
MDL No.: MFCD00036497