A11609.06
A11609.14
A11609.06EA
34.2
EUR
InStock
A11609.06
A11609.14
4-Fluor-o-toluidin ≥97%
4-Fluor-o-toluidin
Purity: 97%
CAS No.: 452-71-1
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 87°(188°F)
Refractive Index: 1.5370
Odour: Characteristic
Boiling Point: 90-92°/16mm
Beilstein Reference: 1931798
UN No.: 2810
MDL No.: MFCD00007832