A11826.14
A11826.06
A11826.14EA
168
EUR
InStock
A11826.14
A11826.06
Methyl-3-(brommethyl)benzoat ≥97%
Methyl-3-(brommethyl)benzoat
Purity: 97%
CAS No.: 1129-28-8
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 43-45°
Boiling Point: 115°/0.8mm
UN No.: 3261
MDL No.: MFCD00051437