A11927.14
A11927.22
A11927.14EA
135
EUR
InStock
A11927.14
A11927.22
tert-Butylethylmalonat 95%
tert-Butylethylmalonat
Purity: 95%
CAS No.: 32864-38-3
Molecular Formula: C9H16O4
Formula Weight: 188.22
Flash Point: 87°(188°F)
Refractive Index: 1.4160
Boiling Point: 98-100°/22mm
Beilstein Reference: 1776681
MDL No.: MFCD00009193