A11943.0B
A11943.0I
A11943.30
A11943.0BEA
227
EUR
InStock
A11943.0B
A11943.0I
A11943.30
3-Aminophenol ≥98%
3-Aminophenol
Purity: 98+%
CAS No.: 591-27-5
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 178°(352°F)
Odour: Characteristic
pH: 6.8
Melting Point: 122-126°
Boiling Point: 164°/11mm
Beilstein Reference: 636059
Merck Reference: 14,460
UN No.: 2512
MDL No.: MFCD00007786