A11949.06
A11949.14
A11949.06EA
93.1
EUR
InStock
A11949.06
A11949.14
2,4-Dimethylimidazol ≥97%
2,4-Dimethylimidazol
Purity: 97%
CAS No.: 930-62-1
Molecular Formula: C5H8N2
Formula Weight: 96.13
Melting Point: 84-88°
Boiling Point: 266°/733mm
UN No.: 3263
MDL No.: MFCD00022365