A11967.14
A11967.22
A11967.14EA
28.3
EUR
InStock
A11967.14
A11967.22
Chlormethylpivalat ≥97%
Chlormethylpivalat
Purity: 97%
CAS No.: 18997-19-8
Molecular Formula: C6H11ClO2
Formula Weight: 150.60
Flash Point: 40°(104°F)
Refractive Index: 1.4170
Boiling Point: 146-148°
Beilstein Reference: 1560838
UN No.: 2924
MDL No.: MFCD00000884