A12024.14
A12024.22
A12024.36
A12024.14EA
43.5
EUR
InStock
A12024.14
A12024.22
A12024.36
Guanin ≥98%
Guanin
Purity: 98%
CAS No.: 73-40-5
Molecular Formula: C5H5N5O
Formula Weight: 151.13
Storage Temperature: 2°C to 8°C
Melting Point: 300°
Beilstein Reference: 9680
Merck Reference: 14,4564
MDL No.: MFCD00071533