A12051.36
A12051.22
A12051.36EA
149
EUR
InStock
A12051.36
A12051.22
Dichlordiphenylsilan ≥97%
Dichloro(diphenyl)silane
Purity: 97%
CAS No.: 80-10-4
Molecular Formula: C12H10Cl2Si
Formula Weight: 253.20
Flash Point: 157°(314°F)
Refractive Index: 1.5790
Odour: Pungent
Melting Point: -22°
Boiling Point: 303-304°
Beilstein Reference: 609882
UN No.: 1769
MDL No.: MFCD00000489