A12058.0B
A12058.22
A12058.30
A12058.0BEA
344
EUR
InStock
A12058.0B
A12058.22
A12058.30
4,4-Dimethoxybutanon ≥90%, tech.
4,4-Dimethoxybutanon
Purity: 90%
CAS No.: 5436-21-5
Molecular Formula: C6H12O3
Formula Weight: 132.16
Flash Point: 57°(134°F)
Refractive Index: 1.4150
Melting Point: -83°
Boiling Point: 170-172°
Beilstein Reference: 1702372
UN No.: 1224
MDL No.: MFCD00008789