A12080.0B
A12080.18
A12080.30
A12080.0BEA
348
EUR
InStock
A12080.0B
A12080.18
A12080.30
Kaliumhexafluorophosphat ≥98%
Potassium hexafluorophosphate
Purity: 98%
CAS No.: 17084-13-8
Molecular Formula: KPF6
Formula Weight: 184.07
Odour: Odourless
pH: 3.0-7.0
Melting Point: 575°
UN No.: 3260
MDL No.: MFCD00011412