A12139.14
A12139.22
A12139.36
A12139.14EA
34.9
EUR
InStock
A12139.14
A12139.22
A12139.36
Phenylacetylen ≥98%
Phenylacetylen
Purity: 98+%
CAS No.: 536-74-3
Molecular Formula: C8H6
Formula Weight: 102.14
Flash Point: 27°(81°F)
Refractive Index: 1.5480
Melting Point: -45°
Boiling Point: 142-144°
Beilstein Reference: 605461
Merck Reference: 14,3861
UN No.: 3295
MDL No.: MFCD00008570