A12161.06
A12161.14
A12161.22
A12161.06EA
28.6
EUR
InStock
A12161.06
A12161.14
A12161.22
2-Chlor-3-nitropyridin ≥99%
2-Chloro-3-nitropyridine
Purity: 99%
CAS No.: 5470-18-8
Molecular Formula: C5H3ClN2O2
Formula Weight: 158.54
Flash Point: 185°(365°F)
Melting Point: 99-103°
Beilstein Reference: 124482
MDL No.: MFCD00006232