A12222.14
A12222.22
A12222.36
A12222.14EA
21.9
EUR
InStock
A12222.14
A12222.22
A12222.36
4'-Bromacetophenon ≥98%
4'-Bromacetophenon
Purity: 98%
CAS No.: 99-90-1
Molecular Formula: C8H7BrO
Formula Weight: 199.05
Flash Point: 110°(230°F)
Melting Point: 49-53°
Boiling Point: 255-256°
Beilstein Reference: 386015
Merck Reference: 14,1403
MDL No.: MFCD00000105