A12362.0B
A12362.0I
A12362.30
A12362.0BEA
65.6
EUR
InStock
A12362.0B
A12362.0I
A12362.30
Diphenylcarbonat ≥99%
Diphenylcarbonat
Purity: 99%
CAS No.: 102-09-0
Molecular Formula: C13H10O3
Formula Weight: 214.22
Flash Point: 168°(334°F)
Odour: Mild
Boiling Point: 301-302°
Beilstein Reference: 1074863
Merck Reference: 14,3602
MDL No.: MFCD00003037