A12377.0F
A12377.AE
A12377.AP
A12377.0FEA
118
EUR
InStock
A12377.0F
A12377.AE
A12377.AP
Buttersäureanhydrid ≥98%
Buttersäureanhydrid
Purity: 98%
CAS No.: 106-31-0
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 88°(190°F)
Refractive Index: 1.4130
Odour: Rancid
Melting Point: -75°
Boiling Point: 198-199°
Beilstein Reference: 1099474
Merck Reference: 14,1594
UN No.: 2739
MDL No.: MFCD00009389