A12386.06
A12386.14
A12386.22
A12386.06EA
34.5
EUR
InStock
A12386.06
A12386.14
A12386.22
L-Histidin-Methylester-dihydrochlorid ≥98%
L-Histidine methyl ester dihydrochloride
Purity: 98+%
CAS No.: 7389-87-9
Molecular Formula: C7H11N3O2.2HCl
Formula Weight: 242.11
Storage Temperature: 2°C to 8°C
Melting Point: 198° dec.
Beilstein Reference: 3572010
Optical Rotation: +10° (c=2 in water)
MDL No.: MFCD00012701