A12404.0B
A12404.0I
A12404.30
A12404.0BEA
63.2
EUR
InStock
A12404.0B
A12404.0I
A12404.30
Benzoesäuremethylester ≥99%
Benzoesäuremethylester
Purity: 99%
CAS No.: 93-58-3
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 82°(179°F)
Physical Form: Liquid
Density: 1.094
Refractive Index: 1.5170
Appearance: Colourless
Melting Point: -12°
Boiling Point: 198-200°
Beilstein Reference: 1072099
Merck Reference: 14,6024
MDL No.: MFCD00008421