A12410.14
A12410.22
A12410.14EA
74.2
EUR
InStock
A12410.14
A12410.22
Iodessigsäure ≥97%
Iodessigsäure
Purity: 97+%
CAS No.: 64-69-7
Molecular Formula: ICH2CO2H
Formula Weight: 185.95
Storage Temperature: 2°C to 8°C
Odour: Pungent
Boiling Point: 208° dec.
Beilstein Reference: 1739079
Merck Reference: 14,5025
UN No.: 2923
MDL No.: MFCD00002685