A12420.06
A12420.14
A12420.22
A12420.06EA
58.8
EUR
InStock
A12420.06
A12420.14
A12420.22
Pentafluorbenzoylchlorid ≥98%
Pentafluorbenzoylchlorid
Purity: 98%
CAS No.: 2251-50-5
Molecular Formula: C7ClF5O
Formula Weight: 230.52
Refractive Index: 1.4535
Boiling Point: 158-159°
UN No.: 3265
MDL No.: MFCD00000657