A12430.03
A12430.06
A12430.14
A12430.03EA
130
EUR
InStock
A12430.03
A12430.06
A12430.14
Cyclobutanol 96%
Cyclobutanol
Purity: 96%
CAS No.: 2919-23-5
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: 21°(69°F)
Refractive Index: 1.4360
Boiling Point: 123-124°
Beilstein Reference: 2035937
UN No.: 1120
MDL No.: MFCD00001318